Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcohol, 91.5%, MilliporeSigma™

Catalog No. 50947692 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-947-692 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-947-692 Supplier MilliporeSigma Supplier No. AX04477

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 64-17-5

Specifications

Packaging Glass Bottle
Flash Point 12°C
Quantity 100 mL
Assay Percent Range Assay (ethanol): 89.5 to 91.5%; Assay (isopropyl alcohol): 4.0 to 6.0%; Assay (methanol): 3.5 to 5.5%
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.