Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcohol, Anhydrous, Contains 0.075% Butylated Hydroxytoluene as a stabilizer, PHOTREX™ Reagent, J.T. Baker™

Catalog No. 02002216 Shop All Avantor Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-002-216 4 L Narrow Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-002-216 Supplier Avantor Supplier No. 922903
Add to Cart
Add to Cart

High purity solvents characterized for use in UV, visible and IR spectrophotometry

    For Use in Liquid Chromatography and Spectrophotometry

Chemical Identifiers

CAS 64-17-5
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name methanol
SMILES CO

Specifications

Quantity 4 L
Flash Point 13.00°C (55.4°F)
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.79kg
Grade Meets ACS reagent chemical requirements
Chemical Name or Material Alcohol, Anhydrous, Contains 0.075% Butylated Hydroxytoluene as a stabilizer
Recommended Storage Store above 11°C (53°F) or product will freeze
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.