Learn More
Electron Microscopy Sciences This low heat flame burner is well suited for microscopy purposes. For use with isopropyl and denatured ethyl alcohol. Made from soda lime glass. Measures 75mm dia x 100 mm high
SDP
Catalog No. NC3782558
Encompass_Preferred
This burner offers a low heat flame, which is well suited for microscopy purposes. For use with isopropyl and denatured ethyl alcohol. The unit is supplied with a ground glass stopper. Made from soda lime glass. Size is 100 ml and measures: 75 mm in diameter and 100 mm high.
Catalog No. NC3782558 Supplier Electron Microscopy Sciences Supplier No. 6421302
This burner offers a low heat flame, which is well suited for microscopy purposes. For use with isopropyl and denatured ethyl alcohol. The unit is supplied with a ground glass stopper. Made from soda lime glass. Size is 100 ml and measures: 75 mm in diameter and 100 mm high.