Learn More
Leica Biosystems ALCOHOL CASE OF 4
Catalog No. NC2061189
Encompass_Preferred
Pure filtered and laboratory tested to ensure a consistent high quality for all formulating, processing and staining procedures in histology and cytology.Available in 100%, 95%, 80%, 70% and Isopropanol.The Isopropanol (Isopropyl Alcohol) is 99% anhydrous and is recommended for small tissue and biopsy processing as well as any other laboratory procedures requiring Isopropanol.
Catalog No. NC2061189 Supplier Leica Biosystems Supplier No. 3803680
Pure filtered and laboratory tested to ensure a consistent high quality for all formulating, processing and staining procedures in histology and cytology.Available in 100%, 95%, 80%, 70% and Isopropanol.The Isopropanol (Isopropyl Alcohol) is 99% anhydrous and is recommended for small tissue and biopsy processing as well as any other laboratory procedures requiring Isopropanol.