Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alcohol Dehydrogenase Antibody (YA1948) | 10 UL
SDP

Catalog No. 5000443587
Encompass_Preferred

The MedChemExpress Alcohol Dehydrogenase Antibody (YA1948) is a rabbit-derived, non-conjugated IgG monoclonal antibody specifically targeting Alcohol Dehydrogenase. This antibody is suitable for use in various research applications, including Western Blot (WB), Immunohistochemistry-Paraffin (IHC-P), and Immunocytochemistry/Immunofluorescence (ICC/IF), providing a valuable tool for studying alcohol metabolism and related physiological processes.

  • Targets alcohol dehydrogenase.
  • Rabbit-derived IgG monoclonal antibody.
  • Non-conjugated.
  • Reacts with human, mouse, and rat samples.
  • Predicted and observed molecular weight for the target protein is 40 kDa.
  • Purified by affinity chromatography.
  • Can be stored at -20°C for 1 year; avoid repeated freeze/thaw cycles.
  • Formulation includes 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% sodium azide, and 0.05% BSA.

Catalog No. 50-004-43587 Supplier Medchemexpress LLC Supplier No. HYP8220310UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The MedChemExpress Alcohol Dehydrogenase Antibody (YA1948) is a rabbit-derived, non-conjugated IgG monoclonal antibody specifically targeting Alcohol Dehydrogenase. This antibody is suitable for use in various research applications, including Western Blot (WB), Immunohistochemistry-Paraffin (IHC-P), and Immunocytochemistry/Immunofluorescence (ICC/IF), providing a valuable tool for studying alcohol metabolism and related physiological processes.

  • Targets alcohol dehydrogenase.
  • Rabbit-derived IgG monoclonal antibody.
  • Non-conjugated.
  • Reacts with human, mouse, and rat samples.
  • Predicted and observed molecular weight for the target protein is 40 kDa.
  • Purified by affinity chromatography.
  • Can be stored at -20°C for 1 year; avoid repeated freeze/thaw cycles.
  • Formulation includes 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% sodium azide, and 0.05% BSA.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.