Learn More
Medchemexpress LLC Alcohol Dehydrogenase Antibody (YA1948) | 50 UL
SDP
Catalog No. 5000444061
Encompass_Preferred
Alcohol Dehydrogenase Antibody (YA1948) is a rabbit-derived and non-conjugated IgG monoclonal antibody, specifically targeting Alcohol Dehydrogenase. This product aids in understanding the critical role of Alcohol Dehydrogenase Protein, a fundamental member of the zinc-containing alcohol dehydrogenase family, in catalyzing the conversion of alcohols to aldehydes or ketones. Its study provides insights into physiological processes, alcohol metabolism, and potential applications in biotechnology and medicine.
  • Targets alcohol dehydrogenase
  • Rabbit-derived IgG monoclonal antibody
  • Non-conjugated
  • Reacts with human, mouse, and rat samples
  • Suitable for Western blot (WB), immunohistochemistry (IHC-P), and immunocytochemistry/immunofluorescence (ICC/IF) applications
  • Liquid appearance
  • Predicted and observed molecular weight of 40 kDa
  • Stored at -20°C for 1 year to avoid repeated freeze/thaw cycles
Catalog No. 50-004-44061 Supplier Medchemexpress LLC Supplier No. HYP8220350UL
Alcohol Dehydrogenase Antibody (YA1948) is a rabbit-derived and non-conjugated IgG monoclonal antibody, specifically targeting Alcohol Dehydrogenase. This product aids in understanding the critical role of Alcohol Dehydrogenase Protein, a fundamental member of the zinc-containing alcohol dehydrogenase family, in catalyzing the conversion of alcohols to aldehydes or ketones. Its study provides insights into physiological processes, alcohol metabolism, and potential applications in biotechnology and medicine.
  • Targets alcohol dehydrogenase
  • Rabbit-derived IgG monoclonal antibody
  • Non-conjugated
  • Reacts with human, mouse, and rat samples
  • Suitable for Western blot (WB), immunohistochemistry (IHC-P), and immunocytochemistry/immunofluorescence (ICC/IF) applications
  • Liquid appearance
  • Predicted and observed molecular weight of 40 kDa
  • Stored at -20°C for 1 year to avoid repeated freeze/thaw cycles