Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alcohol dehydrogenase, Saccharomyces cerevisiae | 9031-72-5 | 141-152 kDa | 25 KU
SDP

Catalog No. 5000398569
Encompass_Preferred

Alcohol dehydrogenase, Saccharomyces cerevisiae is a dimeric protein found in the cytosol of cells. It is the primary enzyme responsible for alcohol consumption in the body, predominantly expressed in the liver, where it plays a role in the detoxification of environmental alcohol. This product is intended for research use only.

  • Soluble in water (1 mg/mL) to form a clear to slightly turbid liquid
  • Appears as a white to off-white solid
  • Sourced from Saccharomyces cerevisiae
  • Activity: 277 U/mg
  • Specific activity: 463 U/mg protein
  • Store at -20°C for 2 years

Catalog No. 50-003-98569 Supplier Medchemexpress LLC Supplier No. 25KUHYP2740
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alcohol dehydrogenase, Saccharomyces cerevisiae is a dimeric protein found in the cytosol of cells. It is the primary enzyme responsible for alcohol consumption in the body, predominantly expressed in the liver, where it plays a role in the detoxification of environmental alcohol. This product is intended for research use only.

  • Soluble in water (1 mg/mL) to form a clear to slightly turbid liquid
  • Appears as a white to off-white solid
  • Sourced from Saccharomyces cerevisiae
  • Activity: 277 U/mg
  • Specific activity: 463 U/mg protein
  • Store at -20°C for 2 years

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.