Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcohol, OmniSolv™ Reagent (HPLC), MilliporeSigma™

Catalog No. MAX04456 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAX04456 1 L
MAX04451 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MAX04456 Supplier MilliporeSigma Supplier No. AX04456
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Components: 90% (w/v) Ethyl Alcohol (CAS Reg. 64-17-5) + 5% (w/v) Methanol (CAS Reg. 67-56-1) + 5% (w/v) 2-Propanol (CAS Reg. 67-63-0)

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Alcohol, OmniSolv™ Reagent
Name Note OmniPure™
CAS 64-17-5, 67-56-1, 67-63-0
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Molecular Formula C2H5OH
Linear Formula CH3CH2OH
Synonym Ethyl Alcohol, Denatured, Ethyl Alcohol, Denatured Alcohol
Formula Weight 46.7
Grade HPLC
Concentration or Composition (by Analyte or Components) Assay
  • Ethanol 89.5-91.5% 2-Propanol 4.0-6.0% Methanol 3.5-5.5% Color (APHA)
  • ≤10 Residue after Evaporation
  • ≤4ppm Titrable Acid
  • ≤0.3μEq/g Titrable Base
  • ≤0.04μEq/g Optical Absorbance at 205nm
  • ≤1.00at 210nm
  • ≤0.65at 220nm
  • ≤0.35at 230nm
  • ≤0.20at 250nm
  • ≤0.04at 270nm
  • ≤0.01at 300nm
  • ≤0.005 Water
  • ≤0.04%Filtered through a 0.2 micron filter To pass test
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.