Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alcohol oxidase | 9073-63-6 | 250 U
SDP

Catalog No. 5000397300
Encompass_Preferred

Alcohol oxidase is a functional enzyme involved in the methanol utilization pathway, isolated from yeast peroxisome. It oxidizes methanol to formaldehyde. One unit is defined as the amount of enzyme that can oxidize 1.0 μmole of methanol to formaldehyde per minute at pH 7.5 at 37°C.

  • Appearance: solid
  • Color: light yellow to yellow
  • EC number: 1.1.3.13
  • Enzyme activity: ≥5 U/mg solid
  • Isoelectric point: 5.4-5.8
  • pH range: 6.5-8.5, with an optimum pH of 7.5
  • Cofactor: FAD, one molecule per subunit
  • Solubility: ≥ 50 mg/mL in H2O
  • Inhibitors include: H2O2, propynal, PCMB, iodoacetate, 1,4-butynediol, sodium acetate, cyclopropanone, 4-hydroxy-2-butynal, diethyl dicarbonate, Hg2+, Ag+, and Cu2+

Catalog No. 50-003-97300 Supplier Medchemexpress LLC Supplier No. 250UHYP2756
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alcohol oxidase is a functional enzyme involved in the methanol utilization pathway, isolated from yeast peroxisome. It oxidizes methanol to formaldehyde. One unit is defined as the amount of enzyme that can oxidize 1.0 μmole of methanol to formaldehyde per minute at pH 7.5 at 37°C.

  • Appearance: solid
  • Color: light yellow to yellow
  • EC number: 1.1.3.13
  • Enzyme activity: ≥5 U/mg solid
  • Isoelectric point: 5.4-5.8
  • pH range: 6.5-8.5, with an optimum pH of 7.5
  • Cofactor: FAD, one molecule per subunit
  • Solubility: ≥ 50 mg/mL in H2O
  • Inhibitors include: H2O2, propynal, PCMB, iodoacetate, 1,4-butynediol, sodium acetate, cyclopropanone, 4-hydroxy-2-butynal, diethyl dicarbonate, Hg2+, Ag+, and Cu2+

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.