Learn More
Sigma Aldrich Fine Chemicals Biosciences Alcohol Oxidase solution f
Catalog No. 502903714
Encompass
Alcohol oxidase (AOX) is a homo-octamer which has a molecular weight of 600-kDa. It has eight flavin adenine dinucleotide (fAD) cofactorsIt belongs to the glucose-methanol-choline (GMC) oxidoreductase family.
Catalog No. 50-290-3714 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A24041KU
Alcohol oxidase (AOX) is a homo-octamer which has a molecular weight of 600-kDa. It has eight flavin adenine dinucleotide (fAD) cofactorsIt belongs to the glucose-methanol-choline (GMC) oxidoreductase family.