Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcohol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019123 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 x 1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9123 5 x 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9123 Supplier MilliporeSigma Supelco Supplier No. PHR13735X1.2ML
Add to Cart
Add to Cart
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Alcohol is a common organic solvent that is widely used in chemistry, biochemistry, and molecular biology. It is also utilized in pharmaceutical and perfume manufacture. Due to its occurrence in alcoholic beverages, it is also widely used in in vivo studies on alcohol-related pathology. Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 64-17-5
Ignition Point 683°F
Vapor Pressure 44.6 mmHg (20°C)
Vapor Density 1.59 (vs. air)
Density 0.789 g/mL (at 25°C)
Flash Point 13°C
Linear Formula CH3 CH2 OH
MDL Number MFCD00003568
Melting Point -114°C
Boiling Point 78°C
Synonym Ethanol; Ethyl alcohol
Type Certified Reference Material
Quantity 5 x 1.2 mL
Formula Weight 46.07
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.