Learn More
Electron Microscopy Sciences WEBCOL™ Alcohol Prep 2 Ply Large
SDP
Catalog No. 50930660
Encompass_Preferred
An economical premium pad material that provides maximum absorbency for scrubbing and cleaning. 70% Isopropyl Alcohol. Sterile, non-woven sponges designed for moderate cleaning.
This item is not returnable. View return policy
Catalog No. 50-930-660 Supplier Electron Microscopy Sciences Supplier No. 7100530
An economical premium pad material that provides maximum absorbency for scrubbing and cleaning. 70% Isopropyl Alcohol. Sterile, non-woven sponges designed for moderate cleaning.