Learn More
Med Vet International Sterile Alcohol Prep Pads 70%, Medium, 200/Box
Catalog No. 502522446
Encompass_Preferred
Before injections, alcohol prep pads are used as a topical antiseptic, anti-infective. The soft, absorbent, non-woven pads are saturated with 70% isopropyl alcohol, USP.
Catalog No. 50-252-2446 Supplier Med Vet International Supplier No. APREP
Before injections, alcohol prep pads are used as a topical antiseptic, anti-infective. The soft, absorbent, non-woven pads are saturated with 70% isopropyl alcohol, USP.