Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Alcohol Prep Pads

Catalog No. 12800201 Shop All Fisherbrand Products
This item is not returnable. View return policy
Catalog No. 12-800-201

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

View Alternate Product

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Alcohol Prep Pads are used to clean an injection site prior to injection to remove bacteria and germs that could be pushed into the skin during the injection process.

  • Easy to use
  • Well absorbed alcohol into the fabric

Specifications

Product Type Alcohol Prep Pad
Color White
Form Pad
Composition 70% IPA, 30% Water
Dimensions (L x W) 3 x 6.5 cm
Weight (Metric) 80 gsm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.