Learn More
Polysciences, Inc. Alcohol, 200 proof, ≥ 99.5%, USP
SDP
Catalog No. NC9258346
Encompass_Preferred
Our histology grade Alcohol Reagent is composed of 90 parts ethyl alcohol, 5 parts methyl alcohol and 5 parts isopropyl alcohol, v/v blend. Preferred mixture for dehydrating tissue samples prior to embedding. Suitable for most histological procedures.
Catalog No. NC9258346 Supplier Polysciences, Inc. Supplier No. 098601
Our histology grade Alcohol Reagent is composed of 90 parts ethyl alcohol, 5 parts methyl alcohol and 5 parts isopropyl alcohol, v/v blend. Preferred mixture for dehydrating tissue samples prior to embedding. Suitable for most histological procedures.