Learn More
E K Industries Reagent Alcohol, 95%, 4 Gal Case
SDP
Catalog No. NC1663357
Encompass_Preferred
Melting point / freezing point ~-114 C, boiling range 78 C - 80 C, Flash point ~13 C
Catalog No. NC1663357 Supplier E K Industries Supplier No. 1201GAL
Melting point / freezing point ~-114 C, boiling range 78 C - 80 C, Flash point ~13 C