Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcohol Reagent ACS, 94 to 96%, MilliporeSigma™

Catalog No. MAX04416 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
4 L
208 L
Packaging:
Glass Bottle
Poly Bottle
Poly-coated Glass Bottle
Steel Barrel
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAX04416 500 mL Glass Bottle
MAX04413 4 L Poly Bottle
50-947-922 4 L Poly-coated Glass Bottle
MAX044120 208 L Steel Barrel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MAX04416 Supplier MilliporeSigma Supplier No. AX04416
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64-17-5 , 7732-18-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
PubChem CID 702
ChEBI CHEBI:16236
SMILES CCO

Specifications

Color Colorless
Quantity 500 mL
Flash Point 12°C
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation ≤0.001%
Assay Percent Range Assay (ethanol and methanol): 94.0 to 96.0% (v/v); Assay (ethanol): 89.0 to 92.0% (v/v); Assay (isopropyl alcohol): 4.0 to 6.0% (v/v); Assay (methanol): 3.5 to 5.5% (v/v)
Linear Formula CH3CH2OH with methanol and isopropyl alcohol
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide
IUPAC Name Ethanol
Percent Purity 94 to 96%
Grade ACS
Chemical Name or Material Alcohol
Show More Show Less
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.