Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC ZEP Alcohol Sanitizer Spray, 55 gal, Liquid, FDA-Regulated

Catalog No. 502840005
Encompass

This Alcohol Sanitizer Spray is an FDA-regulated liquid hand sanitizer formulated for use in food processing facilities. It is supplied in a 55 gallon drum and features an alcohol-based formula, providing effective sanitization.

  • Formulated for use on skin
  • Viscous, colorless, clear liquid
  • Alcohol-like odor
  • Contains at least 65% ethanol
  • Soluble in water
  • Regulated by the FDA as an over-the-counter drug
  • Ships via truck

Catalog No. 50-284-0005 Supplier MSC Supplier No. 94169679
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Alcohol Sanitizer Spray is an FDA-regulated liquid hand sanitizer formulated for use in food processing facilities. It is supplied in a 55 gallon drum and features an alcohol-based formula, providing effective sanitization.

  • Formulated for use on skin
  • Viscous, colorless, clear liquid
  • Alcohol-like odor
  • Contains at least 65% ethanol
  • Soluble in water
  • Regulated by the FDA as an over-the-counter drug
  • Ships via truck

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.