Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcohol SDA 3A 200 Proof, Reagents

Catalog No. 01337527
Change view
Click to view available options
Quantity:
20 L
55 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-527 20 L
01-337-526 55 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-527 Supplier Reagents Holdings Llc Supplier No. C212085020E7
Add to Cart
Add to Cart

Alcohol SDA 3A 200 Proof, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

Synonym Ethanol, Ethyl alcohol, Methylcarbinol

Specifications

Melting Point -114°C
CAS 67-56-1
Color Colorless
Quantity 20 L
Packaging HDPE Winpak
Physical Form Liquid
Specific Gravity 0.7908
Molecular Formula CH4O
Boiling Point 78°C
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
Percent Purity Min. 99.5%
Grade Reagent
Chemical Name or Material Alcohol SDA 3A 200 Proof
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.