Learn More
Scientific Sales Septihol 70% IPA 22oz, 12/cs
SDP
Catalog No. NC1678485
Encompass_Preferred
Septihol Nonsterile Isopropyl Alcohol Solution^ Quick-evaporating 70% (v/v) isopropyl alcohol/30% (v/v) USP purified water blend filtered to 0.22 microns. 651mL bottle with adjustable trigger spray.
Catalog No. NC1678485 Supplier Scientific Sales Supplier No. 625477
Septihol Nonsterile Isopropyl Alcohol Solution^ Quick-evaporating 70% (v/v) isopropyl alcohol/30% (v/v) USP purified water blend filtered to 0.22 microns. 651mL bottle with adjustable trigger spray.