Learn More
Volu Sol Alcoholic Giemsa Stain, 8 oz / 250 mL, Volu-Sol™
Supplier: Volu Sol VAG008
Giemsa Stain, modified. Blood films staining; the nucleus and cytoplasm of white blood cells take on the characteristic blue or pink coloration