Learn More
Cole-Parmer Alconox Alcojet 1404-1 Low Foaming Powdered Detergent; 4 lb Box
Catalog No. NC9336116 Shop All Cole-Parmer Products
Encompass_Preferred
White powder Alconox, Alcojet, and Tergazyme cleaners rinses off completelywith no residue. Alconox cleaner is biodegradable; Alcojet cleaner is low-sudsing; Tergazyme cleaner combines the original Alconox power with protein enzymes. Liquinox cleaner is biodegradableuse to remove soil, grease, blood, chemicals, and solvents. All cleaner are safe, odorless, and FDA compliant.
Catalog No. NC9336116 Supplier Cole-Parmer Supplier No. 1777605
White powder Alconox, Alcojet, and Tergazyme cleaners rinses off completelywith no residue. Alconox cleaner is biodegradable; Alcojet cleaner is low-sudsing; Tergazyme cleaner combines the original Alconox power with protein enzymes. Liquinox cleaner is biodegradableuse to remove soil, grease, blood, chemicals, and solvents. All cleaner are safe, odorless, and FDA compliant.