Learn More
Research Products International Corp Alcojet Low Foaming Powdered Detergent, 4 Pounds
SDP
Catalog No. 50998426
Encompass
Alcojet Low Foaming Powder Detergent is a general purpose nonionic powder for labware, machine washers, manual and ultra-sonic cleaning. Corrosion inhibited for glassware, metals and plastics.
Catalog No. 50-998-426 Supplier Research Products International Corp Supplier No. 114018
Alcojet Low Foaming Powder Detergent is a general purpose nonionic powder for labware, machine washers, manual and ultra-sonic cleaning. Corrosion inhibited for glassware, metals and plastics.