Learn More
Electron Microscopy Sciences Alcojet Low Foaming Powdered Detergent 4 LB
SDP
Supplier:  Electron Microscopy Sciences 6051309
Encompass_Preferred
Non-ionic, low foaming cleaner for use in labware washers, parts washers, power spray systems, manual and ultrasonic cleaning. Corrosion inhibited formula recommended for glassware, metals, plastic, ceramic, porcelain, rubber and fiberglass. Free rinsing for reliable results no interfering residues. USDA authorized. Dilute 1:200 and it has a pH 12.
Catalog No. 50-997-496