Learn More
Sigma Aldrich Fine Chemicals Biosciences AlcojetR detergent pkg of1EA
Catalog No. 502897085
Encompass
Alcojet is a low-foaming powdered detergentConcentrated to save you moneyBiodegradable and readily disposableNon-ionic low foaming cleaner for high performance from your cleaning machinery without damaging washer partsfree rinsing to give you reliable results and no interfering residuesUse to pass your cleaning validation tests for lab accreditation and plant inspection approvalUsed to clean Dissection instruments laboratory ware sampling apparatus plate glass pharmaceutical apparatus cosmetics manufacturing equipment restaurant equipment wine glasses counters floors sidewalks and driveways metal castings forgings and stampings industrial parts tanks and reactors. Authorized by USDA for use in federally inspected meat and poultry plants. Passes inhibitory residue test for water analysis. fDA certified.Used to remove Soil grit grime slime grease tars resins fats oils blood tissue inorganic residues particulates solvents chemicals and
Catalog No. 50-289-7085 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2731981EA
Alcojet is a low-foaming powdered detergentConcentrated to save you moneyBiodegradable and readily disposableNon-ionic low foaming cleaner for high performance from your cleaning machinery without damaging washer partsfree rinsing to give you reliable results and no interfering residuesUse to pass your cleaning validation tests for lab accreditation and plant inspection approvalUsed to clean Dissection instruments laboratory ware sampling apparatus plate glass pharmaceutical apparatus cosmetics manufacturing equipment restaurant equipment wine glasses counters floors sidewalks and driveways metal castings forgings and stampings industrial parts tanks and reactors. Authorized by USDA for use in federally inspected meat and poultry plants. Passes inhibitory residue test for water analysis. fDA certified.Used to remove Soil grit grime slime grease tars resins fats oils blood tissue inorganic residues particulates solvents chemicals and