Learn More
AK GLOBALTECH CORPORATION AlcoMate Revo Breathalyzer, Each, The AlcoMate Revo combines the revolutionary innovation of PRISM technology with state-of-the-art fuel-cell precision.
Catalog No. NC3236531
Encompass_Preferred
Pre-calibrated Replaceable Intelligent Sensor Module (PRISM) TechnologyDOT Approved for Law Enforcement: Accuracy +/-0.005 at 0.10 g/dLFuel-Cell Sensor for Maximum PrecisionUS Coast Guard Approved4-Digit LCD Results DisplayActive Pressure Sensor / Deep Lung Air SamplingCumulative Test CounterLow Battery Indicator / Auto Power-Off
Catalog No. NC3236531 Supplier AK GLOBALTECH CORPORATION Supplier No. REVOEACH
Pre-calibrated Replaceable Intelligent Sensor Module (PRISM) TechnologyDOT Approved for Law Enforcement: Accuracy +/-0.005 at 0.10 g/dLFuel-Cell Sensor for Maximum PrecisionUS Coast Guard Approved4-Digit LCD Results DisplayActive Pressure Sensor / Deep Lung Air SamplingCumulative Test CounterLow Battery Indicator / Auto Power-Off