Learn More
Electron Microscopy Sciences Alconox Anionic Powder Detergent 0.5 OZ
SDP
Catalog No. NC1068095
Encompass_Preferred
The original anionic powder detergent for manual and ultrasonic cleaning. An odorless, high sequestering powder. It helps to decontaminate radioactive surfaces.
Catalog No. NC1068095 Supplier Electron Microscopy Sciences Supplier No. 60512
The original anionic powder detergent for manual and ultrasonic cleaning. An odorless, high sequestering powder. It helps to decontaminate radioactive surfaces.