Learn More
Andwin Scientific Alconox Powder Detergent 1.8kg
SDP
Catalog No. 50997493
Encompass_Preferred
Alconox Powder Detergent 1.8kg concentrate removes soil grit buffing compounds and organic contaminants from glass metal stainless steel porcelain ceramic plastic rubber and fiberglass. Can be used on soft metals. Free rinsing odorless anionic and mild. B
Catalog No. 50-997-493 Supplier Andwin Scientific Supplier No. 60511
Alconox Powder Detergent 1.8kg concentrate removes soil grit buffing compounds and organic contaminants from glass metal stainless steel porcelain ceramic plastic rubber and fiberglass. Can be used on soft metals. Free rinsing odorless anionic and mild. B