Learn More
Electron Microscopy Sciences Det-O-Jet Liquid Detergent 1 QT
SDP
Catalog No. 501898967
Encompass_Preferred
A highly alkaline liquid detergent and wetting agent that is soluble in soft or hard water. It is extremely effective on porcelain, metals, electronic parts, etc. It protects and lubricates the interior working parts of the washer.
Catalog No. 50-189-8967 Supplier Electron Microscopy Sciences Supplier No. 60526
A highly alkaline liquid detergent and wetting agent that is soluble in soft or hard water. It is extremely effective on porcelain, metals, electronic parts, etc. It protects and lubricates the interior working parts of the washer.