Learn More
Hach Company Detergent, Alconox
Catalog No. 501995290 Shop All Hach Company Products
Encompass
Concentrated, anionic detergent for manual and ultrasonic cleaning. Free rinsing to give you reliable results without interfering residues. Ideal for cleaning contaminants from glassware, metals, plastics, ceramic, porcelain, rubber and fiberglass. Dilute 1:100. pH 9.5. Contains phosphate.
Catalog No. 50-199-5290 Supplier Hach Company Supplier No. 2088000
Concentrated, anionic detergent for manual and ultrasonic cleaning. Free rinsing to give you reliable results without interfering residues. Ideal for cleaning contaminants from glassware, metals, plastics, ceramic, porcelain, rubber and fiberglass. Dilute 1:100. pH 9.5. Contains phosphate.