Learn More
Electron Microscopy Sciences Liqui-nox Anionic and Non-Ionic Liquid Detergent 1 QT
SDP
Catalog No. 50997500
Encompass_Preferred
A phosphate free anionic and non-ionic liquid detergent for manual and ultrasonic cleaning. Mild pH.
Catalog No. 50-997-500 Supplier Electron Microscopy Sciences Supplier No. 60516
A phosphate free anionic and non-ionic liquid detergent for manual and ultrasonic cleaning. Mild pH.