Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alconox™ LQ9 Mild Alkaline Liquid Detergent

Catalog No. 16000160 Shop All Alconox Products
Change view
Click to view available options
Volume (English):
1 gal.
5 gal.
15 gal.
55 gal.
Container Type:
Case
Drum
Jerrican
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Container Type
16-000-160 1 gal. Case
16-000-163 55 gal. Drum
16-000-162 15 gal. Drum
16-000-161 5 gal. Jerrican
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 16-000-160 Supplier Alconox™ Supplier No. LQ941
This item is not returnable. View return policy

Concentrated, phosphate-free detergent replaces corrosive acids and hazardous solvents

  • Biodegradable and readily disposable
  • Free rinsing gives reliable results and without interfering residues
  • Use to pass cleaning validation tests for lab accreditation and plant inspection approval
  • Dilute 1:100 for use (10mL in 1L of water)
  • USDA authorized for use in federally inspected meat and poultry plants
  • Passes inhibitory residue test for water analysis
  • Used for phosphate sensitive analysis ware

Used to clean:
Healthcare instruments, laboratory ware, vacuum equipment, tissue culture ware, personal protective equipment, sampling apparatus, catheters, tubing, wine glasses, clean rooms, medical devices, optical parts, electronic components, pharmaceutical apparatus, cosmetics manufacturing equipment, metal castings, forgings and stampings, industrial parts, pipes, tanks and reactors.

Used to remove:
Soil, grit, grime, slime, grease, oils, blood, tissue, particulates, deposits, chemical and solvents.

Surfaces cleaned:
Corrosion inhibited formulation recommended for glass, metal, stainless steel, porcelain, ceramic, plastic, cement and fiberglass. Can be used on soft metals such as copper, aluminum, zinc and magnesium if rinsed promptly. Used for art restoration.

Cleaning method:
Soak, brush, sponge, cloth, ultrasonic, flow through clean-in place.

Shelf life: 2 years from date of manufacture.

Specifications

Product Type Mild Alkaline Liquid Detergent
Form Liquid
pH 8.5
Container Type Case
Content And Storage Store closed and upright in a cool dry place at 15° to 30°C.
For Use With (Application) Cleaning Healthcare instruments, laboratory ware, vacuum equipment, tissue culture ware, personal protective equipment, sampling apparatus
Quantity 4 Pk.
Volume (English) 1 gal.
Volume (Metric) 3.8 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.