Learn More
Research Products International Corp Alconox Powder Detergent, 4 Pounds
SDP
Catalog No. 50212165
Encompass
Concentrated, anionic detergent for manual and ultrasonic cleaning. Free rinsing to give you reliable results without interfering residues. Ideal for cleaning contaminants from glassware, metals, plastics, ceramic, porcelain, rubber and fiberglass. Removes soil, grit, and blood from glass, metal, plastic, rubber and hard surface materials. Cleans effectively in hard, soft, cold or hot water. Excellent replacement for corrosive acids and hazardous solvents.
Catalog No. 50-212-165 Supplier Research Products International Corp Supplier No. 114010
Concentrated, anionic detergent for manual and ultrasonic cleaning. Free rinsing to give you reliable results without interfering residues. Ideal for cleaning contaminants from glassware, metals, plastics, ceramic, porcelain, rubber and fiberglass. Removes soil, grit, and blood from glass, metal, plastic, rubber and hard surface materials. Cleans effectively in hard, soft, cold or hot water. Excellent replacement for corrosive acids and hazardous solvents.