Learn More
Electron Microscopy Sciences Terg-A-Zyme Protease Enzyme 4 LB
SDP
Supplier:  Electron Microscopy Sciences 60520
Encompass_Preferred
Combine Alconox powder with this protease enzyme to effectively remove proteinaceous soils. Ideal for pre-soaking. It also cleans reverse osmosis installations.
Catalog No. 50-997-503