Learn More
Electron Microscopy Sciences Terg-A-Zyme Protease Enzyme 4 LB
SDP
Catalog No. 50997504
Encompass_Preferred
Combine Alconox powder with this protease enzyme to effectively remove proteinaceous soils. Ideal for pre-soaking. It also cleans reverse osmosis installations.
Catalog No. 50-997-504 Supplier Electron Microscopy Sciences Supplier No. 60521
Combine Alconox powder with this protease enzyme to effectively remove proteinaceous soils. Ideal for pre-soaking. It also cleans reverse osmosis installations.