Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alconox™ Tergajet™ Low-Foaming Phosphate-Free Powdered Detergent
GREENER_CHOICE

Catalog No. 16000156 Shop All Alconox Products
Change view
Click to view available options
Volume (English):
4 lb.
25 lb.
50 lb.
300 lb.
Container Type:
Box
Carton
Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Container Type
16-000-156 25 lb. Carton
16-000-159 300 lb. Drum
16-000-155 4 lb. Box
16-000-157 50 lb. Carton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 16-000-156 Supplier Alconox™ Supplier No. 2225
Only null left
Add to Cart
Add to Cart

Nonionic, heavy duty enzyme cleaner for use in labware washers, manual and ultrasonic cleaning

Tergajet is a highly alkaline detergent containing sodium carbonate, silicate of soda, and surfactants.

  • Concentrated, biodegradable and disposable
  • Corrosion inhibited formula
  • Free rinsing, no interfering residues
  • Passes inhibitory residue test
  • Dilute 1:100 for use (10g in 1L of water)
  • Ideal for under-counter and floor standing lab washing machines

Used to clean:
Water testing labware cleaning, phosphate sensitive lab equipment cleaning, hospital instruments, optical components, pharmaceutical apparatus, cosmetics manufacturing equipment, tissue culture ware, plate glass, metal castings, forgings and stampings, industrial parts, animal cages, electronic components, tanks and reactors USDA food contact surface cleaning compliant.

Used to remove:
Soil, radioisotopes, radioactivity, organics, grit, grime, slime, grease, tars, resins, fats, oils, blood, tissue, particulates, inorganic residues, chemical and solvents.

Surfaces cleaned:
Recommended for glass, stainless steel, PTFE, ceramic, cement and alkali resistant plastics and metals. Tergajet is inhibited for Aluminum.

Cleaning method:
Laboratory warewashers, dishwashers, washer-sterilizers, washersanitizers, cart washers, spray washers, electrocleaning, ultrasonic cleaners, clean-in place and manual cleaning if proper skin and eye protection is used.

Shelf life: 2 years from date of manufacture.

Order Info

3.8L (1 gal.) makes up to 98.4L (26 gal.) cleaning solution

Specifications

Product Type Low-Foaming Phosphate-Free Powdered Detergent
Form Powder
pH 11
Appearance White
Container Type Carton
Content And Storage Store closed and upright in a cool dry place at 15° to 30°C.
For Use With (Application) Enzyme cleaner for use in labware washers, manual and ultrasonic cleaning
Volume (English) 25 lb.
Volume (Metric) 11.3 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.