Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alconox™ Tergazyme™ Enzyme-Active Powered Detergent
GREENER_CHOICE

Catalog No. 16000119 Shop All Alconox Products
Change view
Click to view available options
Volume (English):
4 lb.
25 lb.
50 lb.
100 lb.
300 lb.
Container Type:
Box
Carton
Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Container Type
16-000-119 300 lb. Drum
16-000-118 100 lb. Drum
16-000-115 4 lb. Box
16-000-116 25 lb. Carton
16-000-117 50 lb. Carton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 16-000-119 Supplier Alconox™ Supplier No. 1303
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Concentrated, anionic detergent with protease enzyme for manual and ultrasonic cleaning

Tergazyme consists primarily of a homogeneous blend of sodium linear alkylaryl sulfonate, phosphates, carbonates, and protease enzyme.

  • Concentrated, biodegradable and disposable
  • Ideal for removal of proteinaceous soils, tissue, blood and body fluids from glassware, metals, plastic, ceramic, porcelain, rubber and fiberglass
  • Replaces corrosive acids and hazardous solvents
  • Free rinsing gives reliable results without interfering residues
  • Dilute 1:100 for typical use (10g in 1L of water)
  • Phosphate Content: 7.5% (as Phosphorus)
  • Use as cleaning agent in Reverse Osmosis and Ultra-Filtration Systems
  • Use to pass cleaning validation tests for lab accreditation and plant inspection approval
  • USDA authorized for use in federally inspected meat and poultry plants

Used to clean:
Medical instruments/devices, dairy equipment, laboratory ware, reverse osmosis and ultrafiltration membranes and units, sampling apparatus, pharmaceutical apparatus, cosmetics manufacturing equipment, tubing, pipes, optical parts, process equipment, industrial parts, desalination plants, tanks and reactors. Passes inhibitory residue test for water analysis.

Used to remove:
Soil, grit, grime, blood, tissue, grease, fats, oils, proteinaceous soils, dairy proteins, particulates, solvents and bioreactor residue.

Surfaces cleaned:
Corrosion inhibited formulation recommended for glass, metal, stainless steel, porcelain, ceramic, plastic, rubber and fiberglass. Can be used on soft metals such as copper, aluminum, zinc and magnesium if rinsed promptly.

Cleaning method:
Soak, brush, sponge, cloth, ultrasonic, flow through clean-inplace. Will foam—not for spray or machine use.

Shelf life: 2 years from date of manufacture.

Order Info

0.5kg (1 lb.) makes 49.2L (13 gal.) cleaning solution.

Specifications

Product Type Enzyme-Active Powered Detergent
Form Powder
pH 9.5
Appearance White, Cream
Container Type Drum
Content And Storage Store closed and upright in a cool dry place at 15° to 30°C.
For Use With (Application) Remove proteinaceous soils, tissue, blood and body fluids
Volume (English) 300 lb.
Volume (Metric) 136 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.