Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ AlcoSCRUB™ Hand Sanitizer Foam, Pump Bottle

Catalog No. p-6075965 Shop All Epredia Products
Change view
Click to view available options
Volume (Metric):
50 mL
946 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
12-544-104 50 mL
12-544-105 946 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 12-544-104 Supplier Epredia™ Supplier No. 4300224001
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Kill bacteria within 15 seconds using the specially formulated Epredia™ AlcoSCRUB™ Hand Sanitizer Foam.

  • Meets CDC guidelines
  • Formulated with emollients to be gentle on skin
  • 99.99% effective against a broad spectrum of microorganisms

Specifications

Product Type Hand Sanitizer Foam
Description Pump Bottle
Volume (English) 1.7 oz.
Volume (Metric) 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.