Learn More
Electron Microscopy Sciences Alcotabs Effervescent Tablets
SDP
Catalog No. 50365825
Encompass_Preferred
Alconox detergent in an effervescent tablet form for use in syphon type washer rinsers for the cleaning of pipettes and test tubes.
Catalog No. 50-365-825 Supplier Electron Microscopy Sciences Supplier No. 60529
Alconox detergent in an effervescent tablet form for use in syphon type washer rinsers for the cleaning of pipettes and test tubes.