Learn More
Research Products International Corp Alcotabs Tablet Pipet Detergent, 100 Tablets per Bottle
SDP
Catalog No. 50998424
Encompass
Economical Alcotabs are ideal detergent for cleaning glass pipets. For convenient use simply place two tablets in a siphon pipette washer. The tablets slowly dissolve and release effervescent bubbles that provide maximum, cleaning power without leaving residue.
Catalog No. 50-998-424 Supplier Research Products International Corp Supplier No. 114014
Economical Alcotabs are ideal detergent for cleaning glass pipets. For convenient use simply place two tablets in a siphon pipette washer. The tablets slowly dissolve and release effervescent bubbles that provide maximum, cleaning power without leaving residue.