Learn More
Medchemexpress LLC Ald-Ph-amido-C2-PEG3-azide | 1807540-88-0 | 98.9% | 50 MG
SDP
Catalog No. 5000340622
Encompass_Preferred
Ald-Ph-amido-C2-PEG3-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent.
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups
Catalog No. 50-003-40622 Supplier Medchemexpress LLC Supplier No. HY13066750MG
Ald-Ph-amido-C2-PEG3-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent.
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups