Learn More
Medchemexpress LLC Ald-ph-amido-peg4-c2-acid | 1309460-27-2 | 99.1% | 397.42 | C19H27NO8 | 250 MG
SDP
Catalog No. 5000202274
Encompass_Preferred
Ald-Ph-amido-PEG4-C2-acid is a noncleavable polyethylene glycol (PEG4) linker intended for use in antibody-drug conjugate (ADC) research. It provides a stable, non-cleavable connection between an antibody and a payload, and is supplied at high purity to support reproducible bioconjugation.
  • Noncleavable PEG4 linker suitable for ADC development.
  • Terminal carboxylic acid enables amide coupling to payloads.
  • Benzaldehyde moiety enables selective conjugation to hydrazide or aminooxy groups.
  • High purity supports reproducible conjugation results.
  • Molecular weight 397.42 g/mol and formula C19H27NO8.
Catalog No. 50-002-02274 Supplier Medchemexpress LLC Supplier No. HY130097250MG
Ald-Ph-amido-PEG4-C2-acid is a noncleavable polyethylene glycol (PEG4) linker intended for use in antibody-drug conjugate (ADC) research. It provides a stable, non-cleavable connection between an antibody and a payload, and is supplied at high purity to support reproducible bioconjugation.
  • Noncleavable PEG4 linker suitable for ADC development.
  • Terminal carboxylic acid enables amide coupling to payloads.
  • Benzaldehyde moiety enables selective conjugation to hydrazide or aminooxy groups.
  • High purity supports reproducible conjugation results.
  • Molecular weight 397.42 g/mol and formula C19H27NO8.