Learn More
Medchemexpress LLC Ald-Ph-amido-PEG4-C2-acid | 1309460-27-2 | 99.1% | 397.42 g/mol | C19H27NO8 | 500 MG
SDP
Catalog No. 5000202478
Encompass_Preferred
Ald-Ph-amido-PEG4-C2-acid is a noncleavable polyethylene glycol (PEG4) linker designed for use in antibody-drug conjugates (ADCs). It provides a stable, noncleavable connection between an antibody and a cytotoxic payload for research applications. The compound has molecular formula C19H27NO8, molecular weight 397.42 g/mol, and high purity suitable for conjugation workflows.
  • Noncleavable PEG4 linker for ADC development.
  • High purity, 99.1%.
  • CAS number 1309460-27-2 for unambiguous identification.
  • Supplied as 500 MG for laboratory-scale conjugation.
  • Intended for research use only; not for human therapeutic use.
Catalog No. 50-002-02478 Supplier Medchemexpress LLC Supplier No. HY130097500MG
Ald-Ph-amido-PEG4-C2-acid is a noncleavable polyethylene glycol (PEG4) linker designed for use in antibody-drug conjugates (ADCs). It provides a stable, noncleavable connection between an antibody and a cytotoxic payload for research applications. The compound has molecular formula C19H27NO8, molecular weight 397.42 g/mol, and high purity suitable for conjugation workflows.
  • Noncleavable PEG4 linker for ADC development.
  • High purity, 99.1%.
  • CAS number 1309460-27-2 for unambiguous identification.
  • Supplied as 500 MG for laboratory-scale conjugation.
  • Intended for research use only; not for human therapeutic use.