Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ald-Ph-amido-PEG4-C2-NHS ester | 1353011-74-1 | 98.1% | 494.49 g/mol | C23H30N2O10 | 100 MG
SDP

Catalog No. 5000202265
Encompass_Preferred

Ald-Ph-amido-PEG4-C2-NHS ester is a non-cleavable 4-unit polyethylene glycol (PEG) linker functionalized as an N-hydroxysuccinimide (NHS) ester for amine-reactive bioconjugation in antibody-drug conjugate (ADC) development. It is supplied for research use and is typically stored at low temperature.

  • Non-cleavable 4-unit PEG linker for stable conjugation.
  • NHS ester functional group for amine-reactive coupling.
  • Molecular weight 494.49 g/mol.
  • Molecular formula C23H30N2O10.
  • CAS number 1353011-74-1.
  • High purity (≈98.1%) suitable for research applications.
  • Available in multiple package sizes from 50 mg to 10 g.

Catalog No. 50-002-02265 Supplier Medchemexpress LLC Supplier No. HY130101100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ald-Ph-amido-PEG4-C2-NHS ester is a non-cleavable 4-unit polyethylene glycol (PEG) linker functionalized as an N-hydroxysuccinimide (NHS) ester for amine-reactive bioconjugation in antibody-drug conjugate (ADC) development. It is supplied for research use and is typically stored at low temperature.

  • Non-cleavable 4-unit PEG linker for stable conjugation.
  • NHS ester functional group for amine-reactive coupling.
  • Molecular weight 494.49 g/mol.
  • Molecular formula C23H30N2O10.
  • CAS number 1353011-74-1.
  • High purity (≈98.1%) suitable for research applications.
  • Available in multiple package sizes from 50 mg to 10 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.