Learn More
Medchemexpress LLC Ald-Ph-amido-PEG4-C2-NHS ester | 1353011-74-1 | 98.1% | 494.49 g/mol | C23H30N2O10 | 250 MG
SDP
Catalog No. 5000203014
Encompass_Preferred
A noncleavable 4-unit PEG linker bearing an NHS ester reactive group for antibody-agent conjugation. It is supplied for research use and is amine-reactive for coupling payloads to antibody lysines.
  • Noncleavable 4-unit PEG spacer provides hydrophilicity and flexibility.
  • NHS ester functionality for amine-reactive conjugation.
  • High purity suitable for research applications.
  • Soluble in DMSO; protocols provided for in vivo formulations.
  • Stable when stored pure at -20°C.
Catalog No. 50-002-03014 Supplier Medchemexpress LLC Supplier No. HY130101250MG
A noncleavable 4-unit PEG linker bearing an NHS ester reactive group for antibody-agent conjugation. It is supplied for research use and is amine-reactive for coupling payloads to antibody lysines.
  • Noncleavable 4-unit PEG spacer provides hydrophilicity and flexibility.
  • NHS ester functionality for amine-reactive conjugation.
  • High purity suitable for research applications.
  • Soluble in DMSO; protocols provided for in vivo formulations.
  • Stable when stored pure at -20°C.