Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ald-ph-amido-PEG4-C2-NHS ester | 1353011-74-1 | 98.1% | 494.49 g/mol | C23H30N2O10 | 50 MG
SDP

Catalog No. 5000202020
Encompass_Preferred

A noncleavable, four-unit polyethylene glycol (PEG) linker bearing an N-hydroxysuccinimide (NHS) ester functional group designed for antibody-agent conjugation and general bioconjugation applications. The linker provides PEG spacing and a reactive NHS ester for coupling to primary amines; it is supplied as a solid and is intended for research use only.

  • Noncleavable 4-unit PEG linker.
  • NHS ester reactive toward primary amines.
  • Suitable for antibody-agent conjugation and bioconjugation workflows.
  • Molecular weight approximately 494.49 g/mol.
  • CAS number 1353011-74-1.

Catalog No. 50-002-02020 Supplier Medchemexpress LLC Supplier No. HY13010150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A noncleavable, four-unit polyethylene glycol (PEG) linker bearing an N-hydroxysuccinimide (NHS) ester functional group designed for antibody-agent conjugation and general bioconjugation applications. The linker provides PEG spacing and a reactive NHS ester for coupling to primary amines; it is supplied as a solid and is intended for research use only.

  • Noncleavable 4-unit PEG linker.
  • NHS ester reactive toward primary amines.
  • Suitable for antibody-agent conjugation and bioconjugation workflows.
  • Molecular weight approximately 494.49 g/mol.
  • CAS number 1353011-74-1.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.