Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ald-Ph-amido-PEG4-C2-NHS ester | 1353011-74-1 | MFCD20926371 | 98.1% | 494.49 | C23H30N2O10 | 1 G
SDP

Catalog No. 5000203960
Encompass_Preferred

Ald-Ph-amido-PEG4-C2-NHS ester is a noncleavable 4-unit polyethylene glycol (PEG4) NHS ester linker used for antibody-agent conjugation research. It provides an activated N-hydroxysuccinimide (NHS) ester for amine coupling and an aldehyde-phenyl (Ald-Ph) moiety with a two-carbon spacer, suitable for bioconjugation and linker synthesis. Supplied at high reported purity and available in multiple pack sizes.

  • Noncleavable 4-unit PEG linker for antibody-agent conjugation.
  • Activated NHS ester for efficient amine coupling.
  • Aldehyde-phenyl functional group with a C2 spacer.
  • Molecular weight 494.49 g/mol.
  • Purity approximately 98.1% as reported.
  • Available in multiple pack sizes, including 1 G.

Catalog No. 50-002-03960 Supplier Medchemexpress LLC Supplier No. HY1301011G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ald-Ph-amido-PEG4-C2-NHS ester is a noncleavable 4-unit polyethylene glycol (PEG4) NHS ester linker used for antibody-agent conjugation research. It provides an activated N-hydroxysuccinimide (NHS) ester for amine coupling and an aldehyde-phenyl (Ald-Ph) moiety with a two-carbon spacer, suitable for bioconjugation and linker synthesis. Supplied at high reported purity and available in multiple pack sizes.

  • Noncleavable 4-unit PEG linker for antibody-agent conjugation.
  • Activated NHS ester for efficient amine coupling.
  • Aldehyde-phenyl functional group with a C2 spacer.
  • Molecular weight 494.49 g/mol.
  • Purity approximately 98.1% as reported.
  • Available in multiple pack sizes, including 1 G.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.