Learn More
Medchemexpress LLC Ald-Ph-NHS ester | 60444-78-2 | 98.6% | 247.20 g/mol | C12H9NO5 | 1 G
SDP
Catalog No. 5000203156
Encompass_Preferred
Ald-Ph-NHS ester is a non-cleavable linker intended for antibody-drug conjugation (ADC) research. It is supplied as a solid with high purity and multiple pack sizes for laboratory use; intended for research use only.
  • Non-cleavable linker for antibody-drug conjugation applications.
  • Contains an NHS ester functional group for bioconjugation.
  • High purity (98.56%).
  • Available in multiple pack sizes, including 1 G.
  • Solid, white to light yellow appearance, with recommended sealed storage away from moisture.
Catalog No. 50-002-03156 Supplier Medchemexpress LLC Supplier No. HY1301071G
Ald-Ph-NHS ester is a non-cleavable linker intended for antibody-drug conjugation (ADC) research. It is supplied as a solid with high purity and multiple pack sizes for laboratory use; intended for research use only.
  • Non-cleavable linker for antibody-drug conjugation applications.
  • Contains an NHS ester functional group for bioconjugation.
  • High purity (98.56%).
  • Available in multiple pack sizes, including 1 G.
  • Solid, white to light yellow appearance, with recommended sealed storage away from moisture.