Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ald-Ph-NHS ester | 60444-78-2 | 98.6% | 247.20 g/mol | C12H9NO5 | 100 MG
SDP

Catalog No. 5000202175
Encompass_Preferred

Ald-Ph-NHS ester is a non-cleavable linker reagent used for antibody-agent conjugation (ADC) in research applications. The compound features an N-hydroxysuccinimide (NHS) ester for efficient coupling to primary amines, producing stable, non-cleavable conjugates suitable for bioconjugation workflows.

  • Non-cleavable NHS ester linker for ADC conjugation.
  • High purity (98.56%).
  • Soluble in DMSO; may require sonication and anhydrous solvent.
  • CAS number 60444-78-2; molecular weight 247.20 g/mol.
  • Store solid sealed at 4°C; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-02175 Supplier Medchemexpress LLC Supplier No. HY130107100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Ald-Ph-NHS ester is a non-cleavable linker reagent used for antibody-agent conjugation (ADC) in research applications. The compound features an N-hydroxysuccinimide (NHS) ester for efficient coupling to primary amines, producing stable, non-cleavable conjugates suitable for bioconjugation workflows.

  • Non-cleavable NHS ester linker for ADC conjugation.
  • High purity (98.56%).
  • Soluble in DMSO; may require sonication and anhydrous solvent.
  • CAS number 60444-78-2; molecular weight 247.20 g/mol.
  • Store solid sealed at 4°C; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.