Learn More
Medchemexpress LLC Ald-Ph-PEG4-Boc | 1807518-64-4 | C23H35NO8 | 100 MG
SDP
Catalog No. 5000355464
Encompass_Preferred
Ald-Ph-PEG4-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTACs.
  • Facilitates the selective degradation of target proteins.
  • Utilizes the intracellular ubiquitin-proteasome system.
  • Available in various quantities for research needs.
Catalog No. 50-003-55464 Supplier Medchemexpress LLC Supplier No. HY140626100MG
Ald-Ph-PEG4-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTACs.
  • Facilitates the selective degradation of target proteins.
  • Utilizes the intracellular ubiquitin-proteasome system.
  • Available in various quantities for research needs.